Zinkstearat, ZnO 12.5–14 %, Thermo Scientific Chemicals
Zinkstearat, ZnO 12.5–14 %, Thermo Scientific Chemicals
Zinkstearat, ZnO 12.5–14 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinkstearat, ZnO 12.5–14 %, Thermo Scientific Chemicals

CAS: 557-05-1 | C36H70O4Zn | 632.332 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
50 g
250 g
1 kg
5 kg
Katalognummer 033238.A1
auch als 033238-A1 bezeichnet
Preis (EUR)
56,20
Each
Menge:
1 kg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
56,20
Each
Chemikalien-Kennzeichnungen
CAS557-05-1
IUPAC Namezinc(2+) dioctadecanoate
Molecular FormulaC36H70O4Zn
InChI KeyXOOUIPVCVHRTMJ-UHFFFAOYSA-L
SMILES[Zn++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Ash12.5 to 14% ( as ZnO)
Appearance (Color)White to off-white
Assay (unspecified)Softening temperature: 117 to 123 deg C
FormPowder
Zinc stearate is used as a release agent for the manufacturing of rubber, polyurethane and polyester processing. It is a lubricant and thickening agent in cosmetic industry. It plays a vital role as an activator for rubber vulcanization. It acts as a phase transfer catalyst in the saponification of fats due to its lipophilic character. It is an active component in fanning powder, which is utilized to decrease the friction between cards.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only