Natriumtetraphenylborat, ACS, 99.5 % Min., Thermo Scientific Chemicals
Natriumtetraphenylborat, ACS, 99.5 % Min., Thermo Scientific Chemicals
Natriumtetraphenylborat, ACS, 99.5 % Min., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Natriumtetraphenylborat, ACS, 99.5 % Min., Thermo Scientific Chemicals

CAS: 143-66-8 | C24H20BNa | 342.22 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
2 g
10 g
50 g
Katalognummer 036430.18
auch als 036430-18 bezeichnet
Preis (EUR)
235,00
Each
Menge:
50 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
235,00
Each
Chemikalien-Kennzeichnungen
CAS143-66-8
IUPAC Namesodium tetraphenylboranuide
Molecular FormulaC24H20BNa
InChI KeyHFSRCEJMTLMDLI-UHFFFAOYSA-N
SMILES[Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Assay from Supplier's CofA≥99.5% (Gravimetry)
Loss on Drying≤0.5% (constant wt. @105?C)
ClarityPasses test
Appearance (Color)White
FormSolid
Sodium tetraphenylborate is used as a precipitating reagent for potassium, as a protecting agent to protect amino acids. It is used in the preparation of N-acylammonium salts. It finds application as a phenyl donor in palladium-catalyzed cross-coupling reactions to prepare arylalkenes and biaryl compounds. It is used in inorganic and organometallic chemistry due to its solubility in nonpolar solvents. Furthermore, it is involved in the isolation of complexes having dinitrogen ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only