Natriumhexacyanoferrat(II)-Decahydrat, 99 %, Thermo Scientific Chemicals
Natriumhexacyanoferrat(II)-Decahydrat, 99 %, Thermo Scientific Chemicals
Natriumhexacyanoferrat(II)-Decahydrat, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Natriumhexacyanoferrat(II)-Decahydrat, 99 %, Thermo Scientific Chemicals

CAS: 14434-22-1 | C6H20FeN6Na4O10 | 484.06 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
50 g
250 g
Katalognummer 039660.30
auch als 039660-30 bezeichnet
Preis (EUR)
118,00
Each
Menge:
250 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
118,00
Each
Chemikalien-Kennzeichnungen
CAS14434-22-1
Molecular FormulaC6H20FeN6Na4O10
InChI KeyJFSUDVTVQZUDOP-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
Molecular Weight (g/mol)484.06
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Assay (unspecified)98.6% min
Mainly used as important raw material in pharmaceuticals, pigment, tannage, metallurgy and chemical industries. Used in producing iron blue and potassium ferricyanide, surface corrosion protecting for tannage and metal, carburization treating for carbon steel, removing iron in pharmaceutical producing. Used as oxidant, food additive, explosive and chemical reagent. Also used as additive for preventing agglomeration in melt snow in winter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only