Fullerenpulver, 97 % C{70}, Thermo Scientific Chemicals
Fullerenpulver, 97 % C{70}, Thermo Scientific Chemicals
Fullerenpulver, 97 % C{70}, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Fullerenpulver, 97 % C{70}, Thermo Scientific Chemicals

CAS: 115383-22-7 | C70 | 840.77 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
10 mg
50 mg
250 mg
Katalognummer 039720.MD
auch als 039720-MD bezeichnet
Preis (EUR)
537,00
Each
Menge:
250 mg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
537,00
Each
Specifications
Hinweise zum ProzentgehaltC70
CAS115383-22-7
ChEBICHEBI:33195
Chemischer Name oder MaterialFullerene powder
FarbeSchwarz
Mehr anzeigen
Fullerene powder is used in chemical research. It is extensively used for several biomedical applications including the design of high-performance MRI contrast agents, X-Ray imaging contrast agents, photodynamic therapy and drug and gene delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only