Thermo Scientific Chemicals

Selensulfid, 94 %, Thermo Scientific Chemicals

Katalognummer: 042943.22
100 g, Each
Thermo Scientific Chemicals

Selensulfid, 94 %, Thermo Scientific Chemicals

Katalognummer: 042943.22
100 g, Each
Menge
Katalognummer: 042943.22
auch als 042943-22 bezeichnet
Preis (EUR)
Preis 154,00
Online-Preis: 137,06
Ihr Preis
Menge
-

Chemikalien-Kennzeichnungen

CAS
7488-56-4
IUPAC Name
thione
Molecular Formula
S2Se
InChI Key
JNMWHTHYDQTDQZ-UHFFFAOYSA-N
SMILES
S=[Se]=S
Assay (unspecified)
92.6% min

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 3
    Acute Inhalation Toxicity - Dusts and Mists
    Category 3
    Specific target organ toxicity - (repeated exposure)
    Category 2
    Acute aquatic toxicity
    Category 1
    Chronic aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H301 + H331 - Toxic if swallowed or if inhaled

    H373 - May cause damage to organs through prolonged or repeated exposure

    H410 - Very toxic to aquatic life with long lasting effects

    Precautionary Statements

    P260 - Do not breathe dust/fume/gas/mist/vapors/spray

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P311 - Call a POISON CENTER or doctor/physician

    P403 + P233 - Store in a well-ventilated place. Keep container tightly closed

    P405 - Store locked up