2,2-Diphenyl-1-picrylhydrazyl (freie Radikale), 95 %, Thermo Scientific Chemicals
2,2-Diphenyl-1-picrylhydrazyl (freie Radikale), 95 %, Thermo Scientific Chemicals
2,2-Diphenyl-1-picrylhydrazyl (freie Radikale), 95 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2-Diphenyl-1-picrylhydrazyl (freie Radikale), 95 %, Thermo Scientific Chemicals

CAS: 1898-66-4 | C18H12N5O6
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
250 mg
1 g
Katalognummer 044150.MD
auch als 044150-MD bezeichnet
Preis (EUR)
55,65
Exklusiv online
61,90
Ersparnis 6,25 (10%)
Each
Menge:
250 mg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
55,65
Exklusiv online
61,90
Ersparnis 6,25 (10%)
Each
Chemikalien-Kennzeichnungen
CAS1898-66-4
IUPAC Name1,1-diphenyl-2-(2,4,6-trinitrophenyl)hydrazine
Molecular FormulaC18H13N5O6
InChI KeyWCBPJVKVIMMEQC-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(=C(NN(C2=CC=CC=C2)C2=CC=CC=C2)C(=C1)[N+]([O-])=O)[N+]([O-])=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Green to black
Assay (unspecified)≥90.0% ( Elemental analysis or HPLC)
FormPowder
2,2-Diphenyl-1-picrylhydrazyl is used as a sensitive colorimetric free radical scavenger. It serves as a general antioxidant detector. It is utilized to monitor chemical reactions involving radicals as well as a standard of the position and intensity of electron paramagnetic resonance signals. Further, it is employed in the quantitative determination of aliphatic and aromatic thiols by indirect spectroscopy and for photometric determination of tocopherol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only