Thermo Scientific Chemicals

Salpetersäure, 2 N standardisierte Lösung, Thermo Scientific Chemicals

Katalognummer: 044528.AP
500 ml, Each
Thermo Scientific Chemicals

Salpetersäure, 2 N standardisierte Lösung, Thermo Scientific Chemicals

Katalognummer: 044528.AP
500 ml, Each
Menge
Informational:Informatorisch:Für diese Chemikalie benötigen wir möglicherweise zusätzliche Informationen für unsere Unterlagen zur behördlichen und chemischen Konformität. Falls erforderlich, setzen wir uns mit Ihnen in Verbindung, sobald Ihre Bestellung eingegangen ist, um diese Informationen zu erfragen.
Katalognummer: 044528.AP
auch als 044528-AP bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
7697-37-2
IUPAC Name
nitric acid
Molecular Formula
HNO3
InChI Key
GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES
O[N+]([O-])=O
Assay from Supplier's CofA
2.0N
Form
Liquid

Beschreibung

Nitric acid, 2.0N Standardized Solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Substances/mixtures corrosive to metal
    Category 1
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    Physical hazards

    H290 - May be corrosive to metals

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician