Bismut(III)-sulfid, grauer Klumpen, 99.999 % (Metallbasis), Thermo Scientific Chemicals
Bismut(III)-sulfid, grauer Klumpen, 99.999 % (Metallbasis), Thermo Scientific Chemicals
Bismut(III)-sulfid, grauer Klumpen, 99.999 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismut(III)-sulfid, grauer Klumpen, 99.999 % (Metallbasis), Thermo Scientific Chemicals

CAS: 1345-07-9 | Bi2S3 | 514.14 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
50 g
250 g
Katalognummer 045463.18
auch als 045463-18 bezeichnet
Preis (EUR)
356,00
Each
Menge:
50 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
356,00
Each
Chemikalien-Kennzeichnungen
CAS1345-07-9
IUPAC Namedibismuth(3+) trisulfanediide
Molecular FormulaBi2S3
InChI KeyYNRGZHRFBQOYPP-UHFFFAOYSA-N
SMILES[S--].[S--].[S--].[Bi+3].[Bi+3]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Metal Impurities0.001% max.
Bismuth(III) sulfide is mainly used in solar cells, photodiode array and infrared spectroscopy. It is also applied in the manufacture of other bismuth compounds, additives for free-cutting steels and micro-electronics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only