Chrom(II)-chlorid, 99.9 % (Metallbasis), Thermo Scientific Chemicals
Chrom(II)-chlorid, 99.9 % (Metallbasis), Thermo Scientific Chemicals
Chrom(II)-chlorid, 99.9 % (Metallbasis), Thermo Scientific Chemicals
Chrom(II)-chlorid, 99.9 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chrom(II)-chlorid, 99.9 % (Metallbasis), Thermo Scientific Chemicals

CAS: 10049-05-5
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 g
5 g
25 g
Katalognummer 047176.06
auch als 047176-06 bezeichnet
Preis (EUR)
140,00
Each
Zum Warenkorb hinzufügen
Menge:
5 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
140,00
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS10049-05-5
IUPAC Nameλ²-chromium(2+) dichloride
Molecular FormulaCl2Cr
InChI KeyXBWRJSSJWDOUSJ-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Cr++]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Metal Impurities0.1% max.
Chromium(II) chloride is used in the condensation of aldehydes with trisubstituted chloroalkenes as well as in the synthesis of a highly active precatalyst for ethylene oligimerization. Further uses of chromium(II) chloride include the conversion of terminal olefins into trans-iodocyclopropanes (in conjunction with iodoform and tetraethyl ethylenediamine) with excellent results; generating bis(indenyl)chromium dimers and other various related products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only