Ammoniumsulfat, 99.95 % (Metallbasis), Thermo Scientific Chemicals
Ammoniumsulfat, 99.95 % (Metallbasis), Thermo Scientific Chemicals
Ammoniumsulfat, 99.95 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammoniumsulfat, 99.95 % (Metallbasis), Thermo Scientific Chemicals

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 g
500 g
2 kg
Katalognummer 089363.A3
auch als 089363-A3 bezeichnet
Preis (EUR)
349,00
Each
Menge:
2 kg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
349,00
Each
Chemikalien-Kennzeichnungen
CAS7783-20-2
IUPAC Namesulfuric acid diamine
Molecular FormulaH8N2O4S
InChI KeyBFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Metal Impurities0.05% max.
Ammonium sulfate is widely utilized as a fertilizer for alkaline soils. In biochemistry it is used for purification of proteins via ammonium sulfate precipitation method. As a food additive, it serves as an acidity regulator in flours and breads. It is used in the preparation of other ammonium salts, particularly ammonium persulfate. It is applied in several applications as a flame retardant and a wood preservative. It finds use as raw materials for various chemical products such as dyes and pharmaceutical products. A saturated solution of ammonium sulfate in heavy water is employed as an external standard in sulfur-33 NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only