CEDIA™ Buprenorphine II Drugs of Abuse Calibrators and Controls
Das tatsächliche Produkt kann abweichen
Thermo Scientific™

CEDIA™ Buprenorphine II Drugs of Abuse Calibrators and Controls

Monitor assay performance with the Thermo Scientific™ CEDIA™ Buprenorphine II Drugs of Abuse Calibrators and Controls.
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerBeschreibung
10020804CEDIA Buprenorphin-II-Kontrollsatz: Niedrig (7,5 ng/ml), Hoch (12,5 ng/ml)
10020800CEDIA Buprenorphine II Kalibrator 20ng/mL
10020799CEDIA Buprenorphin II 10 ng/ml Grenzwert Kalibrator
10020802CEDIA Buprenorphine II Kalibrator 100ng/mL
10021390CEDIA Negativkalibrator II
10020801CEDIA Buprenorphine II Kalibrator 50ng/mL
Katalognummer 10020804
Preis (EUR)
-
Beschreibung:
CEDIA Buprenorphin-II-Kontrollsatz: Niedrig (7,5 ng/ml), Hoch (12,5 ng/ml)
Achieve qualitative and semi-quantitative test results with the Thermo Scientific™ CEDIA™ Buprenorphine II Calibrators and Controls. The five calibrators, including negative, are used to establish a standard curve that correlates the reaction signals from the analyzer with the calibrators’ respective concentrations (in ng/mL). Two controls, one above and one below the cutoff (10ng/mL), validate the calibration curve. The calibrators and controls also provide a reference point for qualitative (positive or negative results) and semi-quantitative detection.
  • Ensures assay accuracy
  • 18 months shelf life
  • Supports qualitative and semi-quantitative measurements
Für In-vitro-Diagnostik
For In Vitro Diagnostic Use.  
Specifications
KonzentrationLow: 7.5 ng/mL, High: 12.5 ng/mL
BeschreibungCEDIA Buprenorphin-II-Kontrollsatz: Niedrig (7,5 ng/ml), Hoch (12,5 ng/ml)
Menge2 x 5 mL
TypControl
Nachweisbare AnalytenBuprenorphine
ProbentypUrine
Unit SizeEach
Inhalt und Lagerung
2 °C bis 8 °C