2,4,6-Trimethoxybenzaldehyd 98 %, Thermo Scientific Chemicals
2,4,6-Trimethoxybenzaldehyd 98 %, Thermo Scientific Chemicals
2,4,6-Trimethoxybenzaldehyd 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,6-Trimethoxybenzaldehyd 98 %, Thermo Scientific Chemicals

CAS: 830-79-5 | C10H12O4 | 196.2 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
10 g
25 g
250 g
Verpackung:
Glasflasche
Katalognummer 160532500
Preis (EUR)
636,00
Each
Menge:
250 g
Verpackung:
Glasflasche
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
636,00
Each
Chemikalien-Kennzeichnungen
CAS830-79-5
IUPAC Name2,4,6-trimethoxybenzaldehyde
Molecular FormulaC10H12O4
InChI KeyCRBZVDLXAIFERF-UHFFFAOYSA-N
SMILESCOC1=CC(OC)=C(C=O)C(OC)=C1
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Crystalline powder
Melting point118°C to 120°C
Appearance (Color)Beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only