DL-10-Camphersulfonsäure, Natriumsalz, 98 %, Thermo Scientific Chemicals
DL-10-Camphersulfonsäure, Natriumsalz, 98 %, Thermo Scientific Chemicals
DL-10-Camphersulfonsäure, Natriumsalz, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-10-Camphersulfonsäure, Natriumsalz, 98 %, Thermo Scientific Chemicals

CAS: 34850-66-3 | C10H15NaO4S | 254.28 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
500 g
Verpackung:
Alle anzeigen
Kunststoffflasche
Katalognummer 189125000
Preis (EUR)
209,00
Each
Menge:
500 g
Verpackung:
Kunststoffflasche
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
209,00
Each
Chemikalien-Kennzeichnungen
CAS34850-66-3
IUPAC Namesodium {7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl}methanesulfonate
Molecular FormulaC10H15NaO4S
InChI KeyAWMAOFAHBPCBHJ-UHFFFAOYNA-M
SMILES[Na+].CC1(C)C2CCC1(CS([O-])(=O)=O)C(=O)C2
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Titration with NaOH>=97.5 %
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only