Dichlorisocyanursäure Natriumsalz, 98 %, Thermo Scientific Chemicals
Dichlorisocyanursäure Natriumsalz, 98 %, Thermo Scientific Chemicals
Dichlorisocyanursäure Natriumsalz, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichlorisocyanursäure Natriumsalz, 98 %, Thermo Scientific Chemicals

Dichlorisocyanursäure Natriumsalz, >-97,5 % (auf Trockensubstanz), C3HCl2N3NAO3, CAS-Nummer 2893-78-9, Natriumdichlorocyanursäure, 1,3,5-Triazin-2,4,6 1h,3h,5h-Trione, 1,3-Dichloro-, Natriumsalz, 500 g, 98 %, weiß, 220-767-7, 219,95, Gefahr
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 g
500 g
2.5 kg
Katalognummer 204110025
Preis (EUR)
261,65
Online Exclusive
290,00
Ersparnis 28,35 (10%)
Each
Menge:
2.5 kg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
261,65
Online Exclusive
290,00
Ersparnis 28,35 (10%)
Each
Chemikalien-Kennzeichnungen
CAS2893-78-9
IUPAC Namesodium 3,5-dichloro-2,4,6-trioxo-1,3,5-triazinan-1-ide
Molecular FormulaC3Cl2N3NaO3
InChI KeyMSFGZHUJTJBYFA-UHFFFAOYSA-M
SMILES[Na+].ClN1C(=O)[N-]C(=O)N(Cl)C1=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White
Appearance (Form)Crystals or granules
Infrared spectrumConforms
Titration with HClO4>=97.5 % (On dry substance)
Loss on drying=<5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X-Bindungsbildungsreagenz, Oxidationsmittel
RUO – Research Use Only