Thermo Scientific Chemicals

Zirkon(IV)-chlorid, 98 %, wasserfrei, Thermo Scientific Chemicals

Katalognummer: 206410050
5 g, Glasflasche, Each
Thermo Scientific Chemicals

Zirkon(IV)-chlorid, 98 %, wasserfrei, Thermo Scientific Chemicals

Katalognummer: 206410050
5 g, Glasflasche, Each
Menge
Verpackung
Katalognummer: 206410050
Preis (EUR)
Preis 42,80
Online-Preis: 38,52
Ihr Preis
Menge
-

Chemikalien-Kennzeichnungen

CAS
10026-11-6
IUPAC Name
zirconium(4+) tetrachloride
Molecular Formula
Cl4Zr
InChI Key
DUNKXUFBGCUVQW-UHFFFAOYSA-J
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[Zr+4]
Appearance (Form)
Powder
Chromium (Cr)
=<100 ppm
Silicon (Si)
=<150 ppm
Appearance (Color)
White
Aluminium (Al)
=<100 ppm

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Substances/mixtures corrosive to metal
    Category 1
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    Physical Hazards

    H290 - May be corrosive to metals

    EU Specific Hazard Statements

    EUH014 - Reacts violently with water

    EUH071 - Corrosive to the respiratory tract

    Precautionary Statements

    P402 + P404 - Store in a dry place. Store in a closed container

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P284 - Wear respiratory protection

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician