Thermo Scientific Chemicals

Bis(phenylsulfonyl)Sulfid, 96 %, Thermo Scientific Chemicals

Katalognummer: 300830050
5 g, Glasflasche, Each
Thermo Scientific Chemicals

Bis(phenylsulfonyl)Sulfid, 96 %, Thermo Scientific Chemicals

Katalognummer: 300830050
5 g, Glasflasche, Each
Menge
Verpackung

Chemikalien-Kennzeichnungen

CAS
4388-22-1
IUPAC Name
{[(benzenesulfonyl)sulfanyl]sulfonyl}benzene
Molecular Formula
C12H10O4S3
InChI Key
YQUSJUJNDKUWAM-UHFFFAOYSA-N
SMILES
O=S(=O)(SS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Powder or crystals
Melting point
129°C to 134°C
Appearance (Color)
Off-white to pink
Infrared spectrum
Conforms
HPLC
>=95 %

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text