Thermo Scientific Chemicals

Dimethylaluminiumchlorid, 0.9M-Lösung in Heptan, AcroSeal™, Thermo Scientific Chemicals

Katalognummer: 377791000
100 mL, AcroSeal™ Glasflasche, Each
Thermo Scientific Chemicals

Dimethylaluminiumchlorid, 0.9M-Lösung in Heptan, AcroSeal™, Thermo Scientific Chemicals

Katalognummer: 377791000
100 mL, AcroSeal™ Glasflasche, Each
Menge
Verpackung
Katalognummer: 377791000
Preis (EUR)
Preis 85,40
Ihr Preis
Menge
-

Chemikalien-Kennzeichnungen

CAS
1184-58-3
IUPAC Name
chlorodimethylalumane
Molecular Formula
C2H6AlCl
InChI Key
JGHYBJVUQGTEEB-UHFFFAOYSA-M
SMILES
C[Al](C)Cl
Appearance
May develop some turbidity or precipitate
Appearance (Form)
Solution
Titration Argentometric
0.85 to 1.00 M
Appearance (Color)
Clear to slightly hazy colorless

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis-Säure
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 2
    Substances/mixtures which, in contact with water, emit flammable gases
    Category 1
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Specific target organ toxicity - (single exposure)
    Category 3
    Aspiration Toxicity
    Category 1
    Chronic aquatic toxicity
    Category 2
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H304 - May be fatal if swallowed and enters airways

    H314 - Causes severe skin burns and eye damage

    H336 - May cause drowsiness or dizziness

    H411 - Toxic to aquatic life with long lasting effects

    Physical Hazards

    H225 - Highly flammable liquid and vapor

    H260 - In contact with water releases flammable gases which may ignite spontaneously

    EU Specific Hazard Statements

    EUH014 - Reacts violently with water

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P273 - Avoid release to the environment

    P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P331 - Do NOT induce vomiting

    P302 + P335 + P334 - IF ON SKIN: Brush off loose particles from skin. Immerse in cool water