Thermo Scientific Chemicals

4-Aminobenzylamin, 98 %, Thermo Scientific Chemicals

Katalognummer: 391450050
5 g, Glasflasche, Each
Thermo Scientific Chemicals

4-Aminobenzylamin, 98 %, Thermo Scientific Chemicals

Katalognummer: 391450050
5 g, Glasflasche, Each
Menge
Verpackung
Katalognummer: 391450050
Preis (EUR)
Preis 46,80
Online-Preis: 42,12
Ihr Preis
Menge
-

Chemikalien-Kennzeichnungen

CAS
4403-71-8
IUPAC Name
4-(aminomethyl)aniline
Molecular Formula
C7H10N2
InChI Key
BFWYZZPDZZGSLJ-UHFFFAOYSA-N
SMILES
NCC1=CC=C(N)C=C1
Appearance (Form)
Liquid or solid
Refractive index
1.6080 to 1.6120 (20°C, 589 nm)
GC
>=97.5 %
Appearance (Color)
Yellow to light brown
Infrared spectrum
Conforms

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    Precautionary Statements

    P280 - Wear eye protection/ face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician