4,4'-Dimethylbenzophenon, +98 %, Thermo Scientific Chemicals
4,4'-Dimethylbenzophenon, +98 %, Thermo Scientific Chemicals
4,4'-Dimethylbenzophenon, +98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,4'-Dimethylbenzophenon, +98 %, Thermo Scientific Chemicals

CAS: 611-97-2 | C15H14O | 210.28 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 g
10 g
Verpackung:
Glasflasche
Katalognummer 408220010
Preis (EUR)
13,40
Each
Menge:
1 g
Verpackung:
Glasflasche
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
13,40
Each
Chemikalien-Kennzeichnungen
CAS611-97-2
IUPAC Namebis(4-methylphenyl)methanone
Molecular FormulaC15H14O
InChI KeyZWPWLKXZYNXATK-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(=O)C1=CC=C(C)C=C1
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Form)Crystalline powder
GC>=98.0 %
Appearance (Color)White to light brown
Melting point93°C to 97°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only