Borsäure, 99.6 %, ACS-Reagenz, Thermo Scientific Chemicals
Borsäure, 99.6 %, ACS-Reagenz, Thermo Scientific Chemicals
Borsäure, 99.6 %, ACS-Reagenz, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Borsäure, 99.6 %, ACS-Reagenz, Thermo Scientific Chemicals

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
500g
2kg
Verpackung:
Kunststoffflasche
Katalognummer 423480020
Preis (EUR)
107,00
Each
Menge:
2kg
Verpackung:
Kunststoffflasche
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
107,00
Each
Chemikalien-Kennzeichnungen
CAS10043-35-3
IUPAC Nameboric acid
Molecular FormulaBH3O3
InChI KeyKGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILESOB(O)O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration with NaOH>=99.5 %
Heavy metals (as Pb)=<0.001 %
Non-volatiles with MeOH=<0.05 %
Mehr anzeigen
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis-Säure
RUO – Research Use Only