Thermo Scientific Chemicals

Levetiracetam, Thermo Scientific Chemicals

Katalognummer: 462120250
25 g, Each
Thermo Scientific Chemicals

Levetiracetam, Thermo Scientific Chemicals

Katalognummer: 462120250
25 g, Each
Menge
Katalognummer: 462120250
Preis (EUR)
Preis 206,00
Online-Preis: 185,40
Ihr Preis
Menge
-

Chemikalien-Kennzeichnungen

CAS
102767-28-2
Appearance (Color)
White to almost white
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
116°C to 120°C
Specific optical rotation
-76° to -82° (20°C, 589 nm) (2 % in water)

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Serious Eye Damage/Eye Irritation
    Category 2
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 - Harmful if swallowed

    H319 - Causes serious eye irritation

    Precautionary Statements

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P280 - Wear eye protection/ face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P337 + P313 - If eye irritation persists: Get medical advice/attention