Product Image
Thermo Scientific Chemicals

Bambuterol-Hydrochlorid, Thermo Scientific Chemicals

Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerMenge
4649100101 g
4649100505 g
Katalognummer 464910010
Preis (EUR)
93,70
Each
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
93,70
Each
Chemikalien-Kennzeichnungen
CAS81732-46-9
IUPAC Namehydrogen 3-[2-(tert-butylamino)-1-hydroxyethyl]-5-[(dimethylcarbamoyl)oxy]phenyl N,N-dimethylcarbamate chloride
Molecular FormulaC18H30ClN3O5
InChI KeyLBARATORRVNNQM-UHFFFAOYNA-N
SMILES[H+].[Cl-].CN(C)C(=O)OC1=CC(=CC(OC(=O)N(C)C)=C1)C(O)CNC(C)(C)C
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Infrared spectrumConforms
Appearance (Color)White to light yellow
Water=<0.5 %
Titration Argentometric>=97.5 %
Appearance (Form)Crystalline powder
Mehr anzeigen
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only