Heracell™ VIOS 250i AxD Accessories
Heracell™ VIOS 250i AxD Accessories
Heracell™ VIOS 250i AxD Accessories
Thermo Scientific™
Heracell™ VIOS 250i AxD Accessories
The Heracell™ VIOS 250i AxD CO2 Incubator is designed for integration into automated laboratories that leverage a modular design to scale. With the added innovation of automated door control, these incubators feature effective contamination control technologies, in-chamber HEPA filtration, a fully covered water reservoir, and Steri-Run™ proven sterilization cycle.
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerTypEigenschaftenMenge
50171290FußschalterZum barrierefreien/automatisierten Öffnen und Schließen von Türen1
50171289ErsatzschlüsselZum manuellen Öffnen der Klappe zum Heracell-Inkubator™ VIOS 250i AxD CO 2 im Notfall2
Katalognummer 50171290
Preis (EUR)
-
Typ:
Fußschalter
Eigenschaften:
Zum barrierefreien/automatisierten Öffnen und Schließen von Türen
Menge:
1
Automated door opening/closing with remote control pairs the CO2 incubator with robotic cell expansion production processes, increasing efficiency and consistency. Extending ergonomic benefits to humans, this model offers hands-free access when managing large volume culture vessels.
Added Innovation
  • Automated operation of incubator door
  • Simplified door design setup, well suited for robotic interface
  • Automatic stop if an obstruction is encountered
  • Integration-ready for automated production settings that leverage a modular design to scale
Direct Heat Chamber
  • 255 L electropolished stainless steel chamber with compact footprint, easily stackable
  • Adjustable, perforated shelving
THRIVE ™Active Airflow
  • Thermo Scientific™ THRIVE™ active airflow technology gently distributes clean, humidified air throughout the chamber, ensuring homogeneous growth conditions
  • Fast recovery of all parameters ten minutes or less for stable culturing conditions
  • Automatic stop function disables fan operation during door openings to minimize air exchange
ISO 5 HEPA Filtration
  • In-chamber HEPA continuously filters the entire chamber air volume every 60 seconds
  • Achieves ISO 5 clean room-like air quality conditions in five minutes after a 30-second door opening
  • Continuous protection by minimizing ability for contaminants to settle on interior surfaces
Protected Humidity Reservoir
  • Directly heated water reservoir provides stable high relative humidity levels, preventing culture desiccation
  • Condensation free inner chamber, prevents a breeding ground for contaminant
  • 5x faster humidity recovery than traditional water pan designs
In situ sensors and probes
  • Dual temperature probes for over temperature protection with operational back up
  • Single beam, dual wavelength infrared (IR180Si) CO2 sensor
  • O2control ranges 1 to 21% (hypoxic), included only for 50170754, 50170755, and 50170756
Steri-Run, high temperature defection cycle
  • Overnight cycle simplifies cleaning and eliminates the need for separate autoclaving of parts
  • 180°C cycle assures uniform 6-log sterilization on all chamber surfaces
Intuitive, interactive iCAN™ Touchscreen Control
  • Bright, door-mounted display for easy accessibility and viewing
  • Selectable languages simplify operation: English, Spanish, German, French, Italian, Japanese and Mandarin
Specifications
EigenschaftenZum barrierefreien/automatisierten Öffnen und Schließen von Türen
Zur Verwendung mit (Geräte)Heracell™ VIOS 250i AxD CO2 Incubator
Menge1
TypFußschalter
Unit SizeEach