Product Image
Thermo Scientific™
11 mm Crimp Cap for Autosampler Vials
Aluminum crimp closures provide a secure leak-resistant seal.
Have Questions?
KatalognummerMenge
70003-0330301000/Cs.
Katalognummer 70003-033030
Preis (EUR)
-
Menge:
1000/Cs.
Minimize contamination from handling with convenient, pre-assembled Thermo Scientific™ 11 mm Crimp Caps for Autosampler Vials, compatible with most HPLC and GC autosamplers.
  • 11 mm
  • Aluminium
  • Crimp cap
  • Center hole
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
  • Aluminum seals must be applied with a crimping tool
Specifications
FarbeSilver
Außendurchmesser (metrisch)11 mm
Marking SpotNo
MaterialAluminum
VorgeschlitztNo
Menge1000/Cs.
SterilitätNon-sterile
Unit SizeEach