Thermo Scientific Chemicals

1-Brom-4-Fluorbenzol, 99 %, Thermo Scientific Chemicals

Katalognummer: A11630.22
100 g, Each
Thermo Scientific Chemicals

1-Brom-4-Fluorbenzol, 99 %, Thermo Scientific Chemicals

Katalognummer: A11630.22
100 g, Each
Menge
Katalognummer: A11630.22
auch als A11630-22 bezeichnet
Preis (EUR)
Preis 38,50
Online-Preis: 34,65
Ihr Preis
Menge
-

Chemikalien-Kennzeichnungen

CAS
460-00-4
IUPAC Name
1-bromo-4-fluorobenzene
Molecular Formula
C6H4BrF
InChI Key
AITNMTXHTIIIBB-UHFFFAOYSA-N
SMILES
FC1=CC=C(Br)C=C1
Assay (GC)
≥98.5%
Appearance (Color)
Clear, colourless
Refractive Index
1.5250-1.5280
Form
Liquid
Identification (FTIR)
Conforms

Beschreibung

1-Bromo-4-fluorobenzene is used as an intermediate in the synthesis of atypical antipsychotic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Anwendungen
1-Bromo-4-Fluorobenzol wird als Zwischenprodukt bei der Synthese atypischer Antipsychotika verwendet.

Löslichkeit
In Wasser löslich (0.14 g/l).

Hinweise
Von starken Oxidationsmitteln fernhalten. Behälter dicht verschlossen lassen. Kühl und trocken in einem gut versiegelten Behälter lagern.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 3
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Physical Hazards

    H226 - Flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P312 - Call a POISON CENTER or doctor if you feel unwell