N,N'-Diallyl-L-weinsäurediamid, 99 %, Thermo Scientific Chemicals
N,N'-Diallyl-L-weinsäurediamid, 99 %, Thermo Scientific Chemicals
N,N'-Diallyl-L-weinsäurediamid, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N'-Diallyl-L-weinsäurediamid, 99 %, Thermo Scientific Chemicals

CAS: 58477-85-3 | C10H16N2O4 | 228.25 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
50 g
250 g
Katalognummer A12195.30
auch als A12195-30 bezeichnet
Preis (EUR)
245,00
Each
Menge:
250 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
245,00
Each
Chemikalien-Kennzeichnungen
CAS58477-85-3
SpecificationsSpecification SheetDatenblatt
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or flakes
Melting Point (clear melt)181.0-187.0°C
Optical Rotation+115° to +119° (c=3 in methanol)
Assay (GC)≥98.5%
Cross-linking co-monomer for the polymerization of soluble polyacrylamide gels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only