Thermo Scientific Chemicals

2-Acetylthiophen, 99 %, Thermo Scientific Chemicals

Katalognummer: A13062.36
500 g, Each
Thermo Scientific Chemicals

2-Acetylthiophen, 99 %, Thermo Scientific Chemicals

Katalognummer: A13062.36
500 g, Each
Menge
Katalognummer: A13062.36
auch als A13062-36 bezeichnet
Preis (EUR)
Preis 175,00
Online-Preis: 157,50
Ihr Preis
Menge
-

Chemikalien-Kennzeichnungen

CAS
88-15-3
IUPAC Name
1-(thiophen-2-yl)ethan-1-one
Molecular Formula
C6H6OS
InChI Key
WYJOVVXUZNRJQY-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=CS1
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless to yellow to pale brown
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.5640-1.5670 @ 20?C

Beschreibung

2-Acetylthiophene is used as an intermediate in the manufacturing of drugs like Tiamonium Iodide, Suprofan, Stepronin, Tenonitrozole, Namirotene, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Anwendungen
2-Acetylthiophen wird als Zwischenprodukt bei der Herstellung von Arzneimitteln wie Tiamoniumiodid, Suprovan, Stepronin, Tenonitrozol, Namiroten usw. verwendet

Löslichkeit
Unlöslich in Wasser.

Hinweise
Empfindlich gegenüber Licht. Nicht kompatibel mit Oxidationsmitteln.
RUO – Nur für Forschungszwecke

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 3
    Acute dermal toxicity
    Category 3
    Acute Inhalation Toxicity - Dusts and Mists
    Category 3
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H301 + H311 + H331 - Toxic if swallowed, in contact with skin or if inhaled

    EU Specific Hazard Statements

    Combustible liquid

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

    P302 + P350 - IF ON SKIN: Gently wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell