Thermo Scientific Chemicals

4-Fluor-alpha-Methylstyrol, 95 %, Thermo Scientific Chemicals

Katalognummer: A14147.09
10 g, Each
Thermo Scientific Chemicals

4-Fluor-alpha-Methylstyrol, 95 %, Thermo Scientific Chemicals

Katalognummer: A14147.09
10 g, Each
Menge
Katalognummer: A14147.09
auch als A14147-09 bezeichnet
Preis (EUR)
Preis 94,00
Online-Preis: 83,66
Ihr Preis
Menge
-

Chemikalien-Kennzeichnungen

CAS
350-40-3
IUPAC Name
1-fluoro-4-(prop-1-en-2-yl)benzene
Molecular Formula
C9H9F
InChI Key
VIXHMBLBLJSGIB-UHFFFAOYSA-N
SMILES
CC(=C)C1=CC=C(F)C=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥94.0%
Refractive Index
1.5085-1.5145 @ 20?C

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 3
    Label Elements
    Signal Word

    Warning

    Physical Hazards

    H226 - Flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking