Thermo Scientific Chemicals

m-Toluenchlorid, 98 %, Thermo Scientific Chemicals

Katalognummer: A15458.14
25 g, Each
Thermo Scientific Chemicals

m-Toluenchlorid, 98 %, Thermo Scientific Chemicals

Katalognummer: A15458.14
25 g, Each
Menge
Katalognummer: A15458.14
auch als A15458-14 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
1899-93-0
IUPAC Name
3-methylbenzene-1-sulfonyl chloride
Molecular Formula
C7H7ClO2S
InChI Key
KFPMLWUKHQMEBU-UHFFFAOYSA-N
SMILES
CC1=CC(=CC=C1)S(Cl)(=O)=O
Refractive Index
1.5465-1.5515 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Form
Liquid

Beschreibung

Reactant involved in they synthesis of biologically active molecules including small molecule ligands for the Stat3 SH2 domain, aryldisulfonamides with antibacterial activity, quinazoline analogs for the treatment of Gaucher disease, 17β-HSD2 inhibitors for the treatment of osteoporosis, isoindoline-5-propenehydroxamates for use as histone deactylase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Löslichkeit
Es reagiert mit Wasser.

Hinweise
Feuchtigkeitsempfindlich. Unter inertem Gas lagern. Von Feuchtigkeit, Oxidationsmitteln und Basen fernhalten.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    EU Specific Hazard Statements

    EUH029 - Contact with water liberates toxic gas

    Precautionary Statements

    P402 + P404 - Store in a dry place. Store in a closed container

    P280 - Wear eye protection/ face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician