Thermo Scientific Chemicals

L-Alanin, 99 %

Katalognummer: A15804.22
100 g, Each
Thermo Scientific Chemicals

L-Alanin, 99 %

Katalognummer: A15804.22
100 g, Each
Menge
Katalognummer: A15804.22
auch als A15804-22 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
56-41-7
IUPAC Name
(2S)-2-aminopropanoic acid
Molecular Formula
C3H7NO2
InChI Key
QNAYBMKLOCPYGJ-REOHCLBHSA-N
SMILES
C[C@H](N)C(O)=O
Form
Crystals or powder or crystalline powder
Comment
Specification differs for U.S. and non-U.S. material where indicated
Optical Rotation
+13.7 to +15.1 (U.S. specification)
Appearance (Color)
White
Assay from Suppliers CofA
≥98.5% (U.S. specification)

Beschreibung

L-Alanine plays a vital role in glucose-alanine cycle between tissue and liver. It is used for protein construction as well as involved in the metabolism of tryptophan and vitamin pyridoxine. It provides energy for muscles and central nervous system in order to strengthen the immune system. It helps in the metabolism of sugars and organic acids, and enhances the germination rates of Bacillus subtilis spores. It also displays a cholesterol-reducing effect in animals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Löslichkeit
Löslich in Wasser und kaltem Ethanol. Schwer löslich in Pyridin. Unlöslich in Äther und Aceton.

Hinweise
Nicht kompatibel mit starken Oxidationsmitteln.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text