Thermo Scientific Chemicals

3-Chloranisol, 98+%, Thermo Scientific Chemicals

Katalognummer: A17327.18
50 g, Each
Thermo Scientific Chemicals

3-Chloranisol, 98+%, Thermo Scientific Chemicals

Katalognummer: A17327.18
50 g, Each
Menge
Katalognummer: A17327.18
auch als A17327-18 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
2845-89-8
IUPAC Name
1-chloro-3-methoxybenzene
Molecular Formula
C7H7ClO
InChI Key
YUKILTJWFRTXGB-UHFFFAOYSA-N
SMILES
COC1=CC=CC(Cl)=C1
Form
Liquid
Appearance (Color)
Clear colorless
Refractive Index
1.5330-1.5370 @ 20?C
Assay (GC)
≥98.0%

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    EU Specific Hazard Statements

    Combustible liquid