Thermo Scientific Chemicals

4-Iodbenzylbromid, 97 %, Thermo Scientific Chemicals

Katalognummer: A18200.06
5 g, Each
Thermo Scientific Chemicals

4-Iodbenzylbromid, 97 %, Thermo Scientific Chemicals

Katalognummer: A18200.06
5 g, Each
Menge
Katalognummer: A18200.06
auch als A18200-06 bezeichnet
Preis (EUR)
Preis 81,90
Online-Preis: 73,71
Ihr Preis
Menge
-

Chemikalien-Kennzeichnungen

CAS
16004-15-2
IUPAC Name
1-(bromomethyl)-4-iodobenzene
Molecular Formula
C7H6BrI
InChI Key
BQTRMYJYYNQQGK-UHFFFAOYSA-N
SMILES
BrCC1=CC=C(I)C=C1
Appearance (Color)
White to cream to pale yellow
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
75.0-84.0?C
Assay (GC)
≥96.0%

Beschreibung

4-Iodobenzyl bromide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Anwendungen
4-Iodobenzylbromid wird als organisches Zwischenprodukt der chemischen Synthese verwendet.

Löslichkeit
Unlöslich in Wasser.

Hinweise
Lichtempfindlich. Nicht kompatibel mit Oxidationsmitteln, Basen und Aminen.
RUO – Nur für Forschungszwecke

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    Precautionary Statements

    P280 - Wear eye protection/ face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician