L-Aspartic Acid, fiber drum, 50 kg
Ajinomoto Group

L-Aspartic Acid, fiber drum, 50 kg

L-aspartic acid is a non-essential amino acid characterized by its abundant presence in plant proteins and endogenous biosynthesis from oxaloacetic acid within mammalian systems.
Have Questions?
KatalognummerMenge
A2055450 kg
Katalognummer A20554
Preis (EUR)
-
Menge:
50 kg

L-aspartic acid is a non-essential amino acid characterized by its abundant presence in plant proteins and endogenous biosynthesis from oxaloacetic acid within mammalian systems. This amino acid, alongside glutamic acid, serves critical biochemical functions as a general acid in enzyme active sites while maintaining protein solubility and ionic character through its charged side chain properties.

Manufactured by Ajinomoto Health & Nutrition North America, Inc., this high-quality L-aspartic acid product represents advanced amino acid technology contributing to significant developments in nutrition and health applications. The product maintains specifications suitable for diverse scientific and industrial applications, including biotechnology/pharmaceutical formulations, cell culture media preparation, food-grade applications, and intravenous (IV) therapeutic solutions.

Complies with United States Pharmacopeia (USP), European Pharmacopoeia (EP), and Japanese Pharmacopoeia (JP) standards.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS56-84-8
Chemischer Name oder MaterialL-Aspartic acid
VerpackungFiber Drum
GüteEP/JP/USP
Molekulargewicht (g/mol)133.1
Reinheit (%)98.5 to 101.0%
Menge50 kg
Unit SizeEach