L-Arginine HCl
Ajinomoto Group

L-Arginine HCl

L-arginine monohydrochloride is a non-essential amino acid that serves as a unique precursor to nitrogen monoxide (NO), functioning as a vasodilator in biological systems.
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerMenge
A3260250 kg
A325761 kg
Katalognummer A32602
Preis (EUR)
-
Menge:
50 kg

L-arginine monohydrochloride is a non-essential amino acid that serves as a unique precursor to nitrogen monoxide (NO), functioning as a vasodilator in biological systems. This hydrochloride salt form exhibits superior solubility characteristics and reduced organoleptic properties compared to standard arginine, presenting with less characteristic taste and odor. The compound finds primary applications across biotech/pharmaceutical sectors, cell culture media formulations, and intravenous (IV) therapeutic preparations. Ajinomoto Health & Nutrition North America, Inc., manufactures this product as part of their commitment to delivering highest-quality amino acid products while advancing nutritional and health applications through amino acid science.

Complies with United States Pharmacopeia (USP), European Pharmacopoeia (EP), and Japanese Pharmacopoeia (JP) standards.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS1119-34-2
Chemischer Name oder MaterialL-Arginine monohydrochloride
VerpackungPoly Drum
GüteEP/JP/USP
Molekulargewicht (g/mol)210.66
Reinheit (%)98.5 to 101.0%
Menge50 kg
Unit SizeEach