Thermo Scientific Chemicals

Diethyl-(Phthalimidomethyl)phosphonat, 97 %, Thermo Scientific Chemicals

Katalognummer: B21096.14
25 g, Each
Thermo Scientific Chemicals

Diethyl-(Phthalimidomethyl)phosphonat, 97 %, Thermo Scientific Chemicals

Katalognummer: B21096.14
25 g, Each
Menge
Katalognummer: B21096.14
auch als B21096-14 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
33512-26-4
IUPAC Name
diethyl [(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)methyl]phosphonate
Molecular Formula
C13H16NO5P
InChI Key
IUZMHUAHKBHJFY-UHFFFAOYSA-N
SMILES
CCOP(=O)(CN1C(=O)C2=CC=CC=C2C1=O)OCC
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
58.0-68.0?C

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text