Danger
H314 - Causes severe skin burns and eye damage
EUH014 - Reacts violently with water
EUH029 - Contact with water liberates toxic gas
P402 + P404 - Store in a dry place. Store in a closed container
P280 - Wear eye protection/ face protection
P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting
P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing
P310 - Immediately call a POISON CENTER or doctor/physician