Thermo Scientific Chemicals

O-(6-Chlor-1H-Benzotriazol-1-yl)-N,N,N',N'-Tetramethyluronium Hexafluorophosphat, 98+ %, Thermo Scientific Chemicals

Katalognummer: H26402.18
50 g, Each
Thermo Scientific Chemicals

O-(6-Chlor-1H-Benzotriazol-1-yl)-N,N,N',N'-Tetramethyluronium Hexafluorophosphat, 98+ %, Thermo Scientific Chemicals

Katalognummer: H26402.18
50 g, Each
Menge
Katalognummer: H26402.18
auch als H26402-18 bezeichnet
Preis (EUR)
Preis 220,00
Online-Preis: 198,00
Ihr Preis
Menge
-

Chemikalien-Kennzeichnungen

CAS
330645-87-9
IUPAC Name
hexafluoro-λ⁵-phosphanuide; {[(6-chloro-1H-1,2,3-benzotriazol-1-yl)oxy](dimethylamino)methylidene}dimethylazanium
Molecular Formula
C11H15ClF6N5OP
InChI Key
ZHHGTMQHUWDEJF-UHFFFAOYSA-N
SMILES
F[P-](F)(F)(F)(F)F.CN(C)C(ON1N=NC2=CC=C(Cl)C=C12)=[N+](C)C
Loss on Drying
≤0.5%
Form
Powder
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream
Assay (HPLC)
≥98.0%

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P332 + P313 - If skin irritation occurs: Get medical advice/attention

    P337 + P313 - If eye irritation persists: Get medical advice/attention