Thermo Scientific Chemicals

3-Fluorpyridin-2-carboxaldehyd, 98 %, Thermo Scientific Chemicals

Katalognummer: H61902.06
5 g, Each
Thermo Scientific Chemicals

3-Fluorpyridin-2-carboxaldehyd, 98 %, Thermo Scientific Chemicals

Katalognummer: H61902.06
5 g, Each
Menge
Katalognummer: H61902.06
auch als H61902-06 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
31224-43-8
IUPAC Name
3-fluoropyridine-2-carbaldehyde
Molecular Formula
C6H4FNO
InChI Key
OZIMPUNGBUYCSP-UHFFFAOYSA-N
SMILES
FC1=C(C=O)N=CC=C1
Assay (GC)
>97.5%

Beschreibung

3-Fluoropyridine-2-carboxaldehyde is used to prepare taxoids derived from 9β-dihydrobaccatin-9,10-acetals with anti-tumor activities. It is also an intermediate used to synthesize S-3578 derivatives with potent activities against both methicillin-resistant Staphylococcus aureus (MRSA) and Pseudomonas aeruginosa.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Anwendungen
3-Fluorpyridin-2-carboxaldehyd wird zur Herstellung von Taxoiden verwendet, die aus 9β-Dihydrobacccatin-9,10-Acetalen mit Antitumorwirkung abgeleitet werden. Es ist auch ein Zwischenprodukt zur Synthese von S-3578-Derivaten mit starker Wirksamkeit gegen den Methicillin-resistenten Staphylococcus aureus (MRSA) sowie gegen Pseudomonas aeruginosa.

Löslichkeit
Schwer in Wasser löslich.

Hinweise
Kühl und gut belüftet lagern. Behälter dicht verschlossen lassen. Von Oxidationsmitteln entfernt aufbewahren.
RUO – Nur für Forschungszwecke

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water