Thermo Scientific Chemicals

p-Toluenesulfonyl-Semicarbazid, 95 %, Thermo Scientific Chemicals

Katalognummer: H61949.22
100 g, Each
Thermo Scientific Chemicals

p-Toluenesulfonyl-Semicarbazid, 95 %, Thermo Scientific Chemicals

Katalognummer: H61949.22
100 g, Each
Menge
Katalognummer: H61949.22
auch als H61949-22 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
10396-10-8
IUPAC Name
(4-methylbenzenesulfonamido)urea
Molecular Formula
C8H11N3O3S
InChI Key
VRFNYSYURHAPFL-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)S(=O)(=O)NNC(N)=O
Assay (HPLC)
>94.0%

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text