N-Boc-L-Phenylalanin-Methylester, 95 %, Thermo Scientific Chemicals
N-Boc-L-Phenylalanin-Methylester, 95 %, Thermo Scientific Chemicals
N-Boc-L-Phenylalanin-Methylester, 95 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-L-Phenylalanin-Methylester, 95 %, Thermo Scientific Chemicals

CAS: 51987-73-6 | C15H21NO4 | 279.34 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
25 g
Katalognummer H63078.06
auch als H63078-06 bezeichnet
Preis (EUR)
35,40
Each
Menge:
5 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
35,40
Each
Chemikalien-Kennzeichnungen
CAS51987-73-6
SpecificationsSpecification SheetDatenblatt
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only