Thermo Scientific Chemicals

Tricin, 0.5 M-Pufferlösung, pH 8.0

Katalognummer: J62983.AE
100 mL, Each
Thermo Scientific Chemicals

Tricin, 0.5 M-Pufferlösung, pH 8.0

Katalognummer: J62983.AE
100 mL, Each
Menge
Katalognummer: J62983.AE
auch als J62983-AE bezeichnet
Preis (EUR)
Preis 79,00
Online-Preis: 71,10
Ihr Preis
Menge
-

Chemikalien-Kennzeichnungen

CAS
5704-04-1
IUPAC Name
2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}acetic acid
Molecular Formula
C6H13NO5
InChI Key
SEQKRHFRPICQDD-UHFFFAOYSA-N
SMILES
OCC(CO)(CO)NCC(O)=O
Form
Liquid
Concentration
Tricine: 500 mM
pH
8.0 ± 0.2

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    EU Specific Hazard Statements

    EUH210 - Safety data sheet available on request