Tobramycin sulfate
Thermo Scientific Chemicals

Tobramycin sulfate

A water-soluble aminoglycoside antibiotic | CAS: 49842-07-1 | C18H39N5O13S | 565.59 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 mg
1 g
Katalognummer J62995.03
auch als J62995-03 bezeichnet
Preis (EUR)
359,00
Each
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
359,00
Each
Specifications
CAS49842-07-1
Chemischer Name oder MaterialTobramycin sulfate
Formelmasse1425.41
Gesundheitsgefahr 1H302-H315-H319-H335
Gesundheitsgefahr 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Mehr anzeigen
Tobramycin sulfate is used as an antibiotic which is used to treat various types of bacterial infections, particularly Gram-negative infections. It is especially effective against species of Pseudomonas. It is used to inhibit bacterial protein synthesis at the level of 30S (16S rRNA) and 70S ribosomal complex assembly. It is used to treat pseudomonas aeruginosa lung infections and is used in combination with other antibiotics to treat urinary tract infections, gynecologic infections, peritonitis, endocarditis, pneumonia, sepsis, respiratory infections, osteomyelitis and other soft-tissue infections. It is a potential therapy for sinus infections1. Product T1783 has been used to study antibiotic resistance2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tobramycin sulfate is used as an antibiotic which is used to treat various types of bacterial infections, particularly Gram-negative infections. It is especially effective against species of Pseudomonas. It is used to inhibit bacterial protein synthesis at the level of 30S (16S rRNA) and 70S ribosomal complex assembly. It is used to treat pseudomonas aeruginosa lung infections and is used in combination with other antibiotics to treat urinary tract infections, gynecologic infections, peritonitis, endocarditis, pneumonia, sepsis, respiratory infections, osteomyelitis and other soft-tissue infections. It is a potential therapy for sinus infections1. Product T1783 has been used to study antibiotic resistance2.

Solubility
Soluble in water (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Alhariri M & Omri A. Efficacy of liposomal bismuth-ethanedithiol-loaded tobramycin after intratracheal administration in rats with pulmonary Pseudomonas aeruginosa infection. Antimicrob Agents Chemother.2013, 57, (1), 569-78.
  2. Li M & Byron PR Tobramycin disposition in the rat lung following airway administration. J Pharmacol Exp Ther.2013, 347, (2), 318-24.