Econazole nitrate
Econazole nitrate
Econazole nitrate
Thermo Scientific Chemicals

Econazole nitrate

CAS: 24169-02-6 | C18H16Cl3N3O4 | 444.693 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
25 g
Katalognummer J63173.06
auch als J63173-06 bezeichnet
Preis (EUR)
72,20
Each
Menge:
5 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
72,20
Each
Chemikalien-Kennzeichnungen
CAS24169-02-6
IUPAC Name1-{2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl}-1H-imidazole; nitric acid
Molecular FormulaC18H16Cl3N3O4
InChI KeyDDXORDQKGIZAME-UHFFFAOYNA-N
SMILESO[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C=C1
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
FormPowder
Purity≥95.0%
Mass SpecConforms
Appearance (Color)White to off-white
Econazole nitrate is an antifungal medication often used as a cream to treat skin infections like tinea, ringworm and athlete's foot, personal care, cosmetic and pharmaceutical applications. It is used for studies on processes such as platelet serotonin uptake, prostanoid biosynthesis, EDHF-mediated relaxation, and Ca2+ transport pathways in thymic lymphocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only