Thermo Scientific Chemicals

Ubiquitin E1-Inhibitor, PYR-41

Katalognummer: J65388.MC
100 mg, Each
Thermo Scientific Chemicals

Ubiquitin E1-Inhibitor, PYR-41

Katalognummer: J65388.MC
100 mg, Each
Menge
Katalognummer: J65388.MC
auch als J65388-MC bezeichnet
Preis (EUR)
Preis 570,00
Online-Preis: 513,00
Ihr Preis
Menge
-

Chemikalien-Kennzeichnungen

CAS
418805-02-4
IUPAC Name
ethyl 4-{4-[(5-nitrofuran-2-yl)methylidene]-3,5-dioxopyrazolidin-1-yl}benzoate
Molecular Formula
C17H13N3O7
InChI Key
ARGIPZKQJGFSGQ-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=C(C=C1)N1NC(=O)C(=CC2=CC=C(O2)[N+]([O-])=O)C1=O
Appearance (Color)
Dark red to brown
Purity
≥97.0%
Form
Powder

Beschreibung

PYR 41 is an irreversible and cell-permeable UBE1 inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • PYR-41 ist ein zelldurchlässiger Inhibitor von Ubiquitin, das Enzym (E1) aktiviert, mit geringer oder keiner Aktivität gegen E3, E2 oder enzymatischer Caspase-Aktivität
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Skin Sensitization
    Category 1
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 - Harmful if swallowed

    H317 - May cause an allergic skin reaction

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P333 + P313 - If skin irritation or rash occurs: Get medical advice/attention