Cycloheximide, ultrapure, 95%
Cycloheximide, ultrapure, 95%
Cycloheximide, ultrapure, 95%
Thermo Scientific Chemicals

Cycloheximide, ultrapure, 95%

CAS: 66-81-9 | C15H23NO4 | 281.35 g/mol
Have Questions?
KatalognummerMenge
J66901.031 g
Katalognummer J66901.03
Preis (EUR)
82,65
Online exclusive
92,00
Ersparnis 9,35
Each
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
82,65
Online exclusive
92,00
Ersparnis 9,35
Each
Chemikalien-Kennzeichnungen
CAS66-81-9
IUPAC Name4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
Molecular FormulaC15H23NO4
InChI KeyYPHMISFOHDHNIV-QTEFRXOUNA-N
SMILES[H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
FormPowder
Appearance (Color)White to cream
Assay (HPLC)≥ 95.0%
Melting point107°C to 120°C
Potency≥ 900 units/mg (on dry basis)
Mehr anzeigen
A protein synthesis inhibitor in eukaryotic cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only