Thermo Scientific Chemicals

LY 171883, 98%

Katalognummer: J67028.MA
10 mg, Each
Thermo Scientific Chemicals

LY 171883, 98%

Katalognummer: J67028.MA
10 mg, Each
Menge
Katalognummer: J67028.MA
auch als J67028-MA bezeichnet

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 - Harmful if swallowed

    Precautionary Statements

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P312 - Call a POISON CENTER or doctor if you feel unwell