Thermo Scientific Chemicals

Tumor Necrosis Factor-alpha, human

Katalognummer: J67162.IAM
50μg, Each
Thermo Scientific Chemicals

Tumor Necrosis Factor-alpha, human

Katalognummer: J67162.IAM
50μg, Each
Menge
Katalognummer: J67162.IAM
auch als J67162-I& bezeichnet
Preis (EUR)
Preis 221,00
Online-Preis: 196,69
Ihr Preis
Menge
-
Molecular Weight
17.5 ±1.8kDa
Assay from Suppliers CofA
≥95.0% (RP-HPLC)
Form
Powder
Assay from Suppliers CofA
Specific biological assay: ≥2.0x10⁷ IU/mg
Appearance (Color)
White

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text