Thermo Scientific Chemicals

Diethylmaleat, 97 %, Thermo Scientific Chemicals

Katalognummer: L03876.0B
1000 g, Each
Thermo Scientific Chemicals

Diethylmaleat, 97 %, Thermo Scientific Chemicals

Katalognummer: L03876.0B
1000 g, Each
Menge
Katalognummer: L03876.0B
auch als L03876-0B bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
141-05-9
IUPAC Name
1,4-diethyl (2Z)-but-2-enedioate
Molecular Formula
C8H12O4
InChI Key
IEPRKVQEAMIZSS-WAYWQWQTSA-N
SMILES
CCOC(=O)\C=C/C(=O)OCC
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid or viscous liquid
Identification (FTIR)
Conforms
Refractive Index
1.4380-1.4430 @ 20?C
Assay (GC)
≥96.0%

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text

    Sicherheit und Handhabung


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Sensitization
    Category 1
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H317 - May cause an allergic skin reaction

    EU Specific Hazard Statements

    Combustible liquid

    Precautionary Statements

    P280 - Wear eye protection/ face protection