It finds its application in the synthesis of amides using the Ritter reaction with bismuth triflate catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
Findet Anwendung in der Synthese von Amiden unter Verwendung der Ritter-Reaktion mit Wismut Triflate Katalyse.
Löslichkeit
Wasserlöslich.
Hinweise
Von Oxidationsmitteln entfernt aufbewahren Halten Sie den Behälter dicht verschlossen und stellen Sie ihn an einen kühlen, trockenen und gut belüfteten Ort.
RUO – Research Use Only
Allgemeine Referenzen:
- Branislav Jović, et al. FTIR spectroscopic study of hydrogen bonding and solvent induced frequency shifts of N-tert-butylacetamide. J. Mol. Struct.,2013,1044140-143.
- Emmanuel Callens, et al. Synthesis of amides using the Ritter reaction with bismuth triflate catalysis. Tetrahedron Lett.,2006,47(49), 8699-8701.