Thermo Scientific Chemicals

1-Adamantanethanol, 98 %, Thermo Scientific Chemicals

Katalognummer: L09318.03
1 g, Each
Thermo Scientific Chemicals

1-Adamantanethanol, 98 %, Thermo Scientific Chemicals

Katalognummer: L09318.03
1 g, Each
Menge
Katalognummer: L09318.03
auch als L09318-03 bezeichnet
Preis (EUR)
Menge
-

Chemikalien-Kennzeichnungen

CAS
6240-11-5
IUPAC Name
2-(adamantan-1-yl)ethan-1-ol
Molecular Formula
C12H20O
InChI Key
ZBIDZPHRNBZTLT-UHFFFAOYSA-N
SMILES
OCCC12CC3CC(CC(C3)C1)C2
Assay (GC)
>97.5%
Appearance (Color)
White
Form
crystalline powder
Melting Point
70-78?C (≤3?C range)

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Abbildungen

Dokumente und Downloads

Zertifikate

    Häufig gestellte Fragen (FAQ)

    Zitierungen und Referenzen

    Search citations by name, author, journal title or abstract text